Tisk…

Anti-IgG Rabbit Antibody

Dodavatel: Biorbyt
BIRBORB27723-1EA 0 CZK
BIRBORB27723-1 BIRBORB27723-0.5
Anti-IgG Rabbit Antibody
Protilátky
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR