Tisk…

Anti-IgG Rabbit Antibody

Dodavatel: Biorbyt
BIRBORB27749-5EA 0 CZK
BIRBORB27749-5 BIRBORB27749-1
Anti-IgG Rabbit Antibody
Protilátky
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR