Tisk…

Anti-Retinol binding protein 4 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27777-100EA 0 CZK
BIRBORB27777-100
Anti-Retinol binding protein 4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Retinol binding protein 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR