Tisk…

Anti-Mouse FGF21 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB27781-100EA 0 CZK
BIRBORB27781-100
Anti-Mouse FGF21 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Mouse FGF21
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR