Tisk…

Anti-ACTB Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB40724-100EA 0 CZK
BIRBORB40724-100
Anti-ACTB Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:2000

Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR