Tisk…
Anti-ZNHIT2 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB40734-100EA 0 CZK
BIRBORB40734-100
Anti-ZNHIT2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:1000
Type: Primary
Antigen: ZNHIT2 (zinc finger, HIT-type containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat