Tisk…

Anti-ACHE Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB40738-100EA 0 CZK
BIRBORB40738-100
Anti-ACHE Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000

Type: Primary
Antigen: ACHE (acetylcholinesterase (Yt blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR