Tisk…

Anti-FANCC Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB40791-100EA 0 CZK
BIRBORB40791-100
Anti-FANCC Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FANCC (Fanconi anemia, complementation group C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR