Tisk…

Anti-NME5 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB40928-100EA 0 CZK
BIRBORB40928-100
Anti-NME5 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000

Type: Primary
Antigen: NME5 (NME/NM23 family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR