Tisk…

Anti-TMLHE Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB40947-100EA 0 CZK
BIRBORB40947-100
Anti-TMLHE Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-1:1000

Type: Primary
Antigen: TMLHE (trimethyllysine hydroxylase, epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR