Tisk…
Anti-IL12RB2 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB40967-100EA 0 CZK
BIRBORB40967-100
Anti-IL12RB2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000
Type: Primary
Antigen: IL12RB2 (interleukin 12 receptor, beta 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human