Tisk…
Anti-CACNG5 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB40998-100EA 0 CZK
BIRBORB40998-100
Anti-CACNG5 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000
Type: Primary
Antigen: CACNG5 (calcium channel, voltage-dependent, gamma subunit 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human