Tisk…
Anti-GLYCTK Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB41007-100EA 0 CZK
BIRBORB41007-100
Anti-GLYCTK Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000
Type: Primary
Antigen: GLYCTK (glycerate kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human