Tisk…

Anti-KLRK1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41098-100EA 0 CZK
BIRBORB41098-100
Anti-KLRK1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000

Type: Primary
Antigen: KLRK1 (killer cell lectin-like receptor subfamily K, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR