Tisk…

Anti-FBXL12 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41115-100EA 0 CZK
BIRBORB41115-100
Anti-FBXL12 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000

Type: Primary
Antigen: FBXL12 (F-box and leucine-rich repeat protein 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR