Tisk…
Anti-PPP1R16A Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB41156-100EA 0 CZK
BIRBORB41156-100
Anti-PPP1R16A Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000
Type: Primary
Antigen: PPP1R16A (protein phosphatase 1, regulatory subunit 16A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human