Tisk…

Anti-RASA3 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41267-100EA 0 CZK
BIRBORB41267-100
Anti-RASA3 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000

Type: Primary
Antigen: RASA3 (RAS p21 protein activator 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR