Tisk…

Anti-MAF1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41292-100EA 0 CZK
BIRBORB41292-100
Anti-MAF1 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: MAF1 (MAF1 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR