Tisk…
Anti-ZNF707 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB41304-100EA 0 CZK
BIRBORB41304-100
Anti-ZNF707 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500
Type: Primary
Antigen: ZNF707 (zinc finger protein 707)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human