Tisk…
Anti-GIMAP8 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB41477-100EA 0 CZK
BIRBORB41477-100
Anti-GIMAP8 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000
Type: Primary
Antigen: GIMAP8 (GTPase, IMAP family member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human