Tisk…
Anti-DC SIGN Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB41504-100EA 0 CZK
BIRBORB41504-100
Anti-DC SIGN Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200
Type: Primary
Antigen: DC SIGN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human