Tisk…

Anti-TUBB8 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41556-100EA 0 CZK
BIRBORB41556-100
Anti-TUBB8 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: TUBB8 (tubulin, beta 8 class VIII)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR