Tisk…

Anti-FMO2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41579-100EA 0 CZK
BIRBORB41579-100
Anti-FMO2 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500

Type: Primary
Antigen: FMO2 (flavin containing monooxygenase 2 (non-functional))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR