Tisk…

Anti-LAD1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41864-100EA 0 CZK
BIRBORB41864-100
Anti-LAD1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: LAD1 (ladinin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR