Tisk…

Anti-SMAD9 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB41865-100EA 0 CZK
BIRBORB41865-100
Anti-SMAD9 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SMAD9 (SMAD family member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR