Tisk…
Anti-SERPINB6 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB41874-100EA 0 CZK
BIRBORB41874-100
Anti-SERPINB6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: SERPINB6 (serpin peptidase inhibitor, clade B (ovalbumin), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human