Tisk…

Anti-CEACAM5 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB43283-100EA 0 CZK
BIRBORB43283-100
Anti-CEACAM5 Rabbit polyclonal antibody
Protilátky
Type: Primary
Antigen: CEACAM5 (carcinoembryonic antigen-related cell adhesion molecule 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR