Tisk…

Anti-PTGS1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43284-100EA 0 CZK
BIRBORB43284-100
Anti-PTGS1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PTGS1 (prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase and cyclooxygenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR