Tisk…

Anti-SYP Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB43293-100EA 0 CZK
BIRBORB43293-100
Anti-SYP Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: SYP (synaptophysin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR