Tisk…

Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Biorbyt
BIRBORB43298-100EA 0 CZK
BIRBORB43298-100
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR