Tisk…

Anti-IgG Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB43304-100EA 0 CZK
BIRBORB43304-100
Anti-IgG Mouse Monoclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR