Tisk…

Anti-HA tag Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB43333-100EA 0 CZK
BIRBORB43333-100
Anti-HA tag Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR