Tisk…

Anti-EFNB2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43339-100EA 0 CZK
BIRBORB43339-100
Anti-EFNB2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EFNB2 (ephrin-B2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR