Tisk…

Anti-TH Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43341-100EA 0 CZK
BIRBORB43341-100
Anti-TH Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TH (tyrosine hydroxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR