Tisk…

Anti-IGF1R Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43345-100EA 0 CZK
BIRBORB43345-100
Anti-IGF1R Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR