Tisk…

Anti-KDR Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43347-100EA 0 CZK
BIRBORB43347-100
Anti-KDR Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR