Tisk…
Anti-Synapsin Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB43354-100EA 0 CZK
BIRBORB43354-100
Anti-Synapsin Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Synapsin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat