Tisk…

Anti-RUNX1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43431-100EA 0 CZK
BIRBORB43431-100
Anti-RUNX1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RUNX1 (runt-related transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR