Tisk…
Anti-MAP1LC3A Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB43472-100EA 0 CZK
BIRBORB43472-100
Anti-MAP1LC3A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat