Tisk…

Anti-JUP Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43482-100EA 0 CZK
BIRBORB43482-100
Anti-JUP Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: JUP (junction plakoglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR