Tisk…

Anti-E2F2 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB43495-100EA 0 CZK
BIRBORB43495-100
Anti-E2F2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: E2F2 (E2F transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR