Tisk…

Anti-IgG1 (Mouse) None Antibody

Dodavatel: Biorbyt
BIRBORB43531-0.1EA 0 CZK
BIRBORB43531-0.1
Anti-IgG1 (Mouse) None Antibody
Protilátky
Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR