Tisk…

Anti-MYC Mouse Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB43631-0.1EA 0 CZK
BIRBORB43631-0.1
Anti-MYC Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR