Tisk…

Anti-ARAP1 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB43651-0.1EA 0 CZK
BIRBORB43651-0.1
Anti-ARAP1 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ARAP1 (ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR