Tisk…

Anti-Cytokeratin Mouse Monoclonal Antibody (PE (Phycoerythrin))

Dodavatel: Biorbyt
BIRBORB43708-0.1EA 0 CZK
BIRBORB43708-0.1
Anti-Cytokeratin Mouse Monoclonal Antibody (PE (Phycoerythrin))
Protilátky
Type: Primary
Antigen: Cytokeratin
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR