Tisk…

Anti-CRP Mouse Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB43719-0.1EA 0 CZK
BIRBORB43719-0.1
Anti-CRP Mouse Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: CRP (C-reactive protein, pentraxin-related)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR