Tisk…

Anti-CD69 Mouse Monoclonal Antibody (Peridinin Chlorophyll)

Dodavatel: Biorbyt
BIRBORB43785-100EA 0 CZK
BIRBORB43785-100
Anti-CD69 Mouse Monoclonal Antibody (Peridinin Chlorophyll)
Protilátky
Type: Primary
Antigen: CD69 (CD69 molecule)
Clonality: monoclonal
Clone:
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR