Tisk…

Anti-GFAP Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB43801-0.1EA 0 CZK
BIRBORB43801-0.1
Anti-GFAP Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: GFAP (glial fibrillary acidic protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR