Tisk…
Anti-CD52 Mouse Monoclonal Antibody (FITC (Fluorescein))
Dodavatel: Biorbyt
BIRBORB43820-100EA 0 CZK
BIRBORB43820-100
Anti-CD52 Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: CD52 (CD52 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human