Tisk…

Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Dodavatel: Biorbyt
BIRBORB43821-100EA 0 CZK
BIRBORB43821-100
Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Protilátky
Type: Primary
Antigen: CD52 (CD52 molecule)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR